Tropolone tosylate ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 7-Oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzenesulfonate, 7-Oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzenesulphonate, 2-Toluenesulphonyloxytropone, 2-Toluenesulfonyloxytropone, 2-Tosyloxytropone

T1574-5G T1574-25G
TCT1574-005GEA 86.9 CAD
TCT1574-005G TCT1574-25G
Tropolone tosylate ≥99.0% (by GC, titration analysis)
Tropolone tosylate

CAS Number: 38768-08-0
MDL Number: MFCD00671574
Molecular Formula: C14H12O4S
Molecular Weight: 276.31
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 160


Other Product Names / Synonyms: 2-Toluenesulfonyloxytropone, 2-Tosyloxytropone

Formula: C₁₄H₁₂O₄S
MW: 276.31 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00671574
CAS Number: 38768-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf35123070a46fe97043d4720d06d69ac0d7ec0&vid=fcf35123070a46fe97043d4720d06d69ac0d7ec0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR