2,3,6-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T1563-5G T1563-1G
TCT1563-5GEA 348.71 CAD
TCT1563-5G TCT1563-001G
2,3,6-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3,6-Trimethoxybenzoic acid

CAS Number: 60241-74-9
MDL Number: MFCD00092587
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 150

Formula: C₁₀H₁₂O₅
MW: 212.1992 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00092587
CAS Number: 60241-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=711d6ecc2aaf752f620242fcf224bed0ed2bd798&vid=711d6ecc2aaf752f620242fcf224bed0ed2bd798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR