Sodium tetrakis(4-fluorophenyl)borate hydrate ≥98.0% (by HPLC and gravimetric analysis)

Supplier: TCI America

Synonyms: Tetrakis(4-fluorophenyl)boron sodium hydrate

A5131-1G A5131-5G
TCA5131-001GEA 514.04 CAD
TCA5131-001G TCA5131-5G
Sodium tetrakis(4-fluorophenyl)borate hydrate ≥98.0% (by HPLC and gravimetric analysis)
Sodium tetrakis(4-fluorophenyl)borate hydrate

[Precipitation reagent for Cs and titrimetric reagent for nonionic surfactants]
CAS Number: 25776-12-9
MDL Number: MFCD00010732
Molecular Formula: C24H16BF4Na
Molecular Weight: 414.19
Purity/Analysis Method: >98.0% (HPLC,W)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Tetrakis(4-fluorophenyl)boron Sodium

Formula: C₂₄H₁₆BF₄Na
MW: 414.19 g/mol (anhy)
MDL Number: MFCD00010732
CAS Number: 25776-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d20f64b4830120c3fdb9a312389ea8f919ea0fd9&vid=d20f64b4830120c3fdb9a312389ea8f919ea0fd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR