Tetraisopropylthiuram disulfide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Bis(diisopropylthiocarbamoyl) Disulphide, Bis(diisopropylthiocarbamoyl) Disulfide
T1555-25G
T1555-5G
TCT1555-25GEA
978.02
CAD
TCT1555-25G
TCT1555-005G
Tetraisopropylthiuram disulfide ≥98.0% (by total nitrogen basis)
Tetraisopropylthiuram disulfide
CAS Number: 4136-91-8
MDL Number: MFCD00009860
Molecular Formula: C14H28N2S4
Molecular Weight: 352.63
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 115
Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds
Other Product Names / Synonyms: Bis(diisopropylthiocarbamoyl) Disulfide
Formula:
C₁₄H₂₈N₂S₄ MW: 352.63 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00009860 CAS Number: 4136-91-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...