4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
T1542-1G
T1542-5G
TCT1542-1GEA
137.46
CAD
TCT1542-1G
TCT1542-005G
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol ≥97.0% (by GC, titration analysis)
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol
CAS Number: 651-84-3
MDL Number: MFCD00191594
Molecular Formula: C7HF7S
Molecular Weight: 250.13
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 169
Specific Gravity (20/20): 1.66
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Mercapto-alpha,alpha,alpha,2,3,5,6-heptafluorotoluene, 4-Trifluoromethyl-2,3,5,6-tetrafluorothiophenol
Formula:
C₇HF₇S MW: 250.14 g/mol Boiling Pt: 169 °C Density: 1.66 (20 °C) |
MDL Number:
MFCD00191594 CAS Number: 651-84-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...