2,4,6-Tri-tert-butylnitrosobenzene ≥98.0%

Supplier: TCI America

T1534-100MG T1534-1G
TCT1534-100MGEA 170.64 CAD
TCT1534-100MG TCT1534-1G
2,4,6-Tri-tert-butylnitrosobenzene ≥98.0%
2,4,6-Tri-tert-butylnitrosobenzene

CAS Number: 24973-59-9
MDL Number: MFCD00008817
Molecular Formula: C18H29NO
Molecular Weight: 275.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Pale Green
Melting point (°C): 168
Storage Temperature: 0-10°C

Formula: C₁₈H₂₉NO
Melting Pt: 168 °C
MDL Number: MFCD00008817
CAS Number: 24973-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc2f91253e01d047c6cdc01a682547f1bf279f5e&vid=cc2f91253e01d047c6cdc01a682547f1bf279f5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR