Tris(8-quinolinolato)aluminum ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
8-Quinolinol Aluminum Complex, Alq3, Aluminum Tris(8-hydroxyquinolinate), Aluminum triquinolin-8-olate
CAS Number: 2085-33-8
MDL Number: MFCD00191693
Molecular Formula: C27H18AlN3O3
Molecular Weight: 459.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow
Melting point (°C): 418
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: Aluminum Tris(8-hydroxyquinolinate), 8-Quinolinol Aluminum Complex, Alq3
Formula:
C₂₇H₁₈AlN₃O₃ MW: 459.44 g/mol Melting Pt: 418 °C |
MDL Number:
MFCD00191693 CAS Number: 2085-33-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...