(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0%

Supplier: TCI America

Synonyms: (S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol, (S)-(+)-alpha-(Trifluoromethyl)-9-anthracenemethanol

T1521-100MG T1521-1G
TCT1521-100MGEA 131.79 CAD
TCT1521-100MG TCT1521-1G
(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0%
(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol

[e.e. Determination Reagent by NMR]
CAS Number: 60646-30-2
MDL Number: MFCD00062968
Molecular Formula: C16H11F3O
Molecular Weight: 276.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 135
Specific rotation [a]20/D: 30.5 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(+)-1-(9-Anthryl)-2,2,2-trifluoroethanol, (S)-(+)-alpha-(Trifluoromethyl)-9-anthracenemethanol

Formula: C₁₆H₁₁F₃O
MW: 276.26 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00062968
CAS Number: 60646-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd12043d6628eea1b62945f719adfbea96999624&vid=fd12043d6628eea1b62945f719adfbea96999624&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR