(2-Trimethylsilylethyl)triphenylphosphonium Iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1510-5G T1510-1G
TCT1510-005GEA 581.28 CAD
TCT1510-005G 76220-498
(2-Trimethylsilylethyl)triphenylphosphonium Iodide ≥98.0% (by HPLC, titration analysis)
(2-Trimethylsilylethyl)triphenylphosphonium Iodide

CAS Number: 63922-84-9
MDL Number: MFCD00191591
Molecular Formula: C23H28IPSi
Molecular Weight: 490.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 117


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Triphenyl(2-trimethylsilylethyl)phosphonium Iodide

Formula: C₂₃H₂₈IPSi
Melting Pt: 117 °C
MDL Number: MFCD00191591
CAS Number: 63922-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a301742ba4dbd1ce85769465e3fa4a6f91a5ecfe&vid=a301742ba4dbd1ce85769465e3fa4a6f91a5ecfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR