Potassium-2,4,5-trichlorophenoxyacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1509-500G T1509-25G
TCT1509-500GEA 309.68 CAD
TCT1509-500G TCT1509-025G
Potassium-2,4,5-trichlorophenoxyacetate ≥98.0% (by titrimetric analysis)
Potassium-2,4,5-trichlorophenoxyacetate

CAS Number: 37785-57-2
MDL Number: MFCD00191590
Molecular Formula: C8H5Cl3O3
Molecular Weight: 293.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 2,4,5-Trichlorophenoxyacetic Acid Potassium Salt, 2,4,5-T Potassium Salt

Formula: C₈H₅Cl₃O₃
MDL Number: MFCD00191590
CAS Number: 37785-57-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR