(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol ≥98.0%
Supplier: TCI America
T1499-1G
T1499-5G
TCT1499-001GEA
133.83
CAD
TCT1499-001G
TCT1499-5G
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol ≥98.0%
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol
CAS Number: 95061-46-4
MDL Number: MFCD00134424
Molecular Formula: C20H18O2
Molecular Weight: 290.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 126
Specific rotation [a]20/D: 220 deg (C=1, EtOH)
Formula:
C₂₀H₁₈O₂ Melting Pt: 126 °C |
MDL Number:
MFCD00134424 CAS Number: 95061-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...