(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol ≥98.0%

Supplier: TCI America

T1491-1G T1491-100MG
TCT1491-1GEA 909.13 CAD
TCT1491-1G TCT1491-100MG
(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol ≥98.0%
(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol

CAS Number: 34281-92-0
MDL Number: MFCD00066432
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 105
Melting point (°C): 65
Specific rotation [a]20/D: 58 deg (C=1, MeOH)

Formula: C₁₂H₁₆O
Boiling Pt: 105 °C
Melting Pt: 65 °C
MDL Number: MFCD00066432
CAS Number: 34281-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b994cd54f42bd5d21b7b91e40531413037e64f&vid=f9b994cd54f42bd5d21b7b91e40531413037e64f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR