N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥97.0%
Supplier: TCI America
Synonyms:
TPEN, N1,N1,N2,N2-Tetrakis(pyridin-2-ylmethyl)ethane-1,2-diamine, Tetrakis-(2-pyridylmethyl)ethylenediamine
T1487-1G
T1487-5G
TCT1487-001GEA
338.12
CAD
TCT1487-001G
TCT1487-5G
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥97.0%
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine
CAS Number: 16858-02-9
MDL Number: MFCD00036918
Molecular Formula: C26H28N6
Molecular Weight: 424.55
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 112
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: TPEN
Formula:
C₂₆H₂₈N₆ MW: 424.55 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD00036918 CAS Number: 16858-02-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...