1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose ≥97.0%
Supplier: TCI America
Synonyms:
(2S,3S,4R,5R,6R)-6-(Acetoxymethyl)-3-hydroxytetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose
T1459-1G
T1459-5G
TCT1459-001GEA
275.62
CAD
TCT1459-001G
TCT1459-005G
1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose ≥97.0%
1,3,4,6-Tetra-O-acetyl-beta-D-mannopyranose
CAS Number: 18968-05-3
MDL Number: MFCD00012354
Molecular Formula: C14H20O10
Molecular Weight: 348.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 162
Specific rotation [a]20/D: -22.5 deg (C=1.4, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₁₄H₂₀O₁₀ MW: 348.30 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD00012354 CAS Number: 18968-05-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...