2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride ≥98.0%

Supplier: TCI America

Synonyms: SEM-triphenylphosphonium Chloride, Triphenyl((2-(trimethylsilyl)ethoxy)methyl)phosphonium chloride

T1458-25G T1458-5G
TCT1458-25GEA 755.4 CAD
TCT1458-25G TCT1458-005G
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride ≥98.0%
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride

CAS Number: 82495-75-8
MDL Number: MFCD00043159
Molecular Formula: C24H30ClOPSi
Molecular Weight: 429.01
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 165


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: SEM-triphenylphosphonium Chloride

Formula: C₂₄H₃₀ClOPSi
MW: 429.01 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00043159
CAS Number: 82495-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96fa6e35579f92a795fe15943b6f5087e8dbb50a&vid=96fa6e35579f92a795fe15943b6f5087e8dbb50a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR