2,3,5,6-Tetramethyl-m-phenylenediamine ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
3,6-Diaminodurene, 2,3,5,6-Tetramethylbenzene-1,3-diamine, 3,6-Diamino-1,2,4,5-tetramethylbenzene, 1,4-Diamino-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-phenylenediamine, 2,3,5,6-Tetramethyl-1,3-phenylenediamine
CAS Number: 3102-87-2
MDL Number: MFCD00007905
Molecular Formula: C10H16N2
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 152
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 3,6-Diaminodurene, 3,6-Diamino-1,2,4,5-tetramethylbenzene
Formula:
C₁₀H₁₆N₂ MW: 164.25 g/mol Boiling Pt: 316 Melting Pt: 152 °C |
MDL Number:
MFCD00007905 CAS Number: 3102-87-2 EINECS: 221-457-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...