2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T1431-25G
T1431-5G
TCT1431-25GEA
1009.62
CAD
TCT1431-25G
TCT1431-005G
2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one ≥98.0% (by titrimetric analysis)
2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one
CAS Number: 95111-49-2
MDL Number: MFCD00067043
Molecular Formula: C7H3Br4NO3
Molecular Weight: 468.72
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Storage Temperature: <0°C
Formula:
C₇H₃Br₄NO₃ |
MDL Number:
MFCD00067043 CAS Number: 95111-49-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...