2-Amino-4,5-dimethylthiazole hydrochloride 98,0

Supplier: TCI America

Synonyms: 4,5-Dimethylthiazol-2-amine hydrochloride

A0292-5G A0292-1G
TCA0292-5GEA 213.3 CAD
TCA0292-5G TCA0292-001G
2-Amino-4,5-dimethylthiazole hydrochloride 98,0
2-Amino-4,5-dimethylthiazole hydrochloride

CAS Number: 71574-33-9
MDL Number: MFCD00012711
Molecular Formula: C5H8N2S
Molecular Weight: 164.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₉ClN₂S
MW: 164.66 g/mol
MDL Number: MFCD00012711
CAS Number: 71574-33-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da7494bff960e5c0d9ff64bcf29cb2b9b2ce5663&vid=da7494bff960e5c0d9ff64bcf29cb2b9b2ce5663&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR