1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole ≥98.0%

Supplier: TCI America

Synonyms: 1-((2,4,6-Triisopropylphenyl)sulfonyl)-1H-imidazole, 1-((2,4,6-Triisopropylphenyl)sulphonyl)-1H-imidazole

T1410-25G T1410-5G
TCT1410-025GEA 712.58 CAD
TCT1410-025G TCT1410-005G
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole ≥98.0%
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole

CAS Number: 50257-40-4
MDL Number: MFCD00005283
Molecular Formula: C18H26N2O2S
Molecular Weight: 334.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 119

Formula: C₁₈H₂₆N₂O₂S
MW: 334.48 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00005283
CAS Number: 50257-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d4d22de0dfc3e969ae5ae5650df662e726fb008&vid=3d4d22de0dfc3e969ae5ae5650df662e726fb008&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR