D-(-)-Luciferin ≥98.0% (by HPLC)

Supplier: TCI America

A5030-10MG A5030-50MG
TCA5030-10MGEA 308.1 CAD
TCA5030-10MG TCA5030-50MG
D-(-)-Luciferin ≥98.0% (by HPLC)
D(-)-Luciferin

[Chemiluminescence Reagent]
CAS Number: 2591-17-5
MDL Number: MFCD00042929
Molecular Formula: C11H8N2O3S2
Molecular Weight: 280.32
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 186
Specific rotation [a]20/D: -33 deg (C=1, DMF)
Lambda max.: 360 nm (H2O)Storage Temperature: <0°C


Product Categories: Bioscience, Luciferase Substrate, Analytical Science, Firefly Luciferin (Chemiluminescence Reagents)
Other Product Names / Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic Acid

Formula: C₁₁H₈N₂O₃S₂
Melting Pt: 186 °C
MDL Number: MFCD00042929
CAS Number: 2591-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=946085306c15b3c0d8f722e420e882f0f7d953f0&vid=2da55005546610695c29e87f604496e32c50dff8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR