Tribehenin ≥90.0% (by titrimetric analysis)
Supplier: TCI America
T1391-5G
T1391-1G
TCT1391-005GEA
360.24
CAD
TCT1391-005G
TCT1391-001G
Tribehenin ≥90.0% (by titrimetric analysis)
Tribehenin
CAS Number: 18641-57-1
MDL Number: MFCD00056274
Molecular Formula: C69H134O6
Molecular Weight: 1059.83
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White
Melting point (°C): 83
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
Formula:
C₆₉H₁₃₄O₆ Melting Pt: 83 °C |
MDL Number:
MFCD00056274 CAS Number: 18641-57-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...