Tricyclo[6.2.1.0(2,7)]undeca-4-ene ≥96.0%

Supplier: TCI America

T1384-25ML T1384-500ML
TCT1384-025MLEA 94.8 CAD
TCT1384-025ML TCT1384-500ML
Tricyclo[6.2.1.0(2,7)]undeca-4-ene ≥96.0%
Tricyclo[6.2.1.0(2,7)]undeca-4-ene

CAS Number: 91465-71-3
MDL Number: MFCD00153968
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 209
Specific Gravity (20/20): 0.96

Formula: C₁₁H₁₆
Boiling Pt: 209 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00153968
CAS Number: 91465-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55cad6c860bf48bb1e667b556fbf4b61328c5065&vid=55cad6c860bf48bb1e667b556fbf4b61328c5065&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR