Tetrafluoroisophthalic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1374-5G T1374-1G
TCT1374-5GEA 412.38 CAD
TCT1374-5G TCT1374-001G
Tetrafluoroisophthalic acid ≥98.0% (by titrimetric analysis)
Tetrafluoroisophthalic acid

CAS Number: 1551-39-9
MDL Number: MFCD00042379
Molecular Formula: C8H2F4O4
Molecular Weight: 238.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 211

Formula: C₈H₂F₄O₄
Melting Pt: 211 °C
MDL Number: MFCD00042379
CAS Number: 1551-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed1fc30d3fdb5cc48924a673f63e6afe255bcdf8&vid=ed1fc30d3fdb5cc48924a673f63e6afe255bcdf8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR