1H-1,2,3-Triazolo[4,5-b]pyridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1367-1G T1367-5G
TCT1367-001GEA 77.42 CAD
TCT1367-001G TCT1367-5G
1H-1,2,3-Triazolo[4,5-b]pyridine ≥98.0% (by HPLC, titration analysis)
1H-1,2,3-Triazolo[4,5-b]pyridine

CAS Number: 273-34-7
MDL Number: MFCD00134183
Molecular Formula: C5H4N4
Molecular Weight: 120.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 207


Other Product Names / Synonyms: 4-Azabenzotriazole

Formula: C₅H₄N₄
Melting Pt: 207 °C
MDL Number: MFCD00134183
CAS Number: 273-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a8688d8580ab0dac2aec781084ccd8c51a68643&vid=9a8688d8580ab0dac2aec781084ccd8c51a68643&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR