(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0%

Supplier: TCI America

T1366-1G T1366-25G
TCT1366-001GEA 74.26 CAD
TCT1366-001G TCT1366-25G
(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0%
(3S)-(-)-3-(Trifluoroacetamido)pyrrolidine hydrochloride

CAS Number: 132883-43-3
MDL Number: MFCD00143195
Molecular Formula: C6H9F3N2O
Molecular Weight: 218.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 233
Specific rotation [a]20/D: -27 deg (C=1, H2O)

Formula: C₆H₉F₃N₂O
MW: 218.60 g/mol
Melting Pt: 233 °C
MDL Number: MFCD00143195
CAS Number: 132883-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7df9c2c8562bf4198ad590585f25a9278018b90a&vid=7df9c2c8562bf4198ad590585f25a9278018b90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR