Trimethylamine-N-oxide, anhydrous ≥95.0% (by titrimetric analysis)
Supplier: TCI America
T1362-1G
T1362-5G
TCT1362-1GEA
69.09
CAD
TCT1362-1G
TCT1362-5G
Trimethylamine-N-oxide, anhydrous ≥95.0% (by titrimetric analysis)
Trimethylamine N-oxide
CAS Number: 1184-78-7
MDL Number: MFCD00002048
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: TMANO
Formula:
C₃H₉NO Melting Pt: 222 °C |
MDL Number:
MFCD00002048 CAS Number: 1184-78-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...