Tris(4-bromophenyl)amine ≥98.0%
Supplier: TCI America
T1361-25G
T1361-5G
TCT1361-025GEA
230.68
CAD
TCT1361-025G
TCT1361-5G
Tris(4-bromophenyl)amine ≥98.0%
Tris(4-bromophenyl)amine
CAS Number: 4316-58-9
MDL Number: MFCD00009665
Molecular Formula: C18H12Br3N
Molecular Weight: 482.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 146
Formula:
C₁₈H₁₂Br₃N MW: 482.01 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD00009665 CAS Number: 4316-58-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...