5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

T1360-100MG T1360-1G
TCT1360-100MGEA 37.92 CAD
TCT1360-100MG TCT1360-1G
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin ≥97.0% (by HPLC)
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin

CAS Number: 22112-78-3
MDL Number: MFCD00012072
Molecular Formula: C48H38N4O4
Molecular Weight: 734.86
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: Deep Blue


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

Formula: C₄₈H₃₈N₄O₄
MW: 734.86 g/mol
MDL Number: MFCD00012072
CAS Number: 22112-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5b6c6b827075bf7f5bdb2bc4fce05e01de46523&vid=f5b6c6b827075bf7f5bdb2bc4fce05e01de46523&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR