Polyethylene glycol dimethacrylate, M.W. n=∼4 stabilised

Supplier: TCI America

Synonyms: PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate

T1352-25G T1352-500G
TCT1352-25GEA 37.92 CAD
TCT1352-25G TCT1352-500G
Polyethylene glycol dimethacrylate, M.W. n=∼4 stabilised
Polyethylene glycol dimethacrylate

(stabilized with MEHQ)
CAS Number: 25852-47-5
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 192
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers

Formula: (C₂H₄O)nC₈H₁₀O₃
Density: 1.08 (20 °C)
Flash Pt: 192 °C
MDL Number: MFCD00014932
CAS Number: 25852-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1dda7bb291f5e1c38c90a5d1bea489a87b3882&vid=8a1dda7bb291f5e1c38c90a5d1bea489a87b3882&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR