Tricyclo[5.2.1.0(2,6)]-8-decanone ≥95.0%

Supplier: TCI America

T1321-25ML T1321-500ML
TCT1321-025MLEA 96.38 CAD
TCT1321-025ML TCT1321-500ML
Tricyclo[5.2.1.0(2,6)]-8-decanone ≥95.0%
Tricyclo[5.2.1.0(2,6)]-8-decanone

CAS Number: 13380-94-4
MDL Number: MFCD00168034
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 132
Flash Point (°C): 102
Specific Gravity (20/20): 1.05

Formula: C₁₀H₁₄O
Boiling Pt: 132 °C
Density: 1.05 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00168034
CAS Number: 13380-94-4
EINECS: 236-458-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69dbec0cf515de01f6fef807b731a08e082b082&vid=d69dbec0cf515de01f6fef807b731a08e082b082&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR