Tetramethyldiphosphine disulfide

Supplier: TCI America

Synonyms: Tetramethylbiphosphine disulphide, Tetramethylbiphosphine disulfide

T1308-5G T1308-1G
TCT1308-005GEA 423.44 CAD
TCT1308-005G TCT1308-1G
Tetramethyldiphosphine disulfide
Tetramethyldiphosphine disulfide

CAS Number: 3676-97-9
MDL Number: MFCD00014459
Molecular Formula: C4H12P2S2
Molecular Weight: 186.21
Form: Crystal
Color: White
Melting point (°C): 226

Formula: C₄H₁₂P₂S₂
MW: 186.21 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00014459
CAS Number: 3676-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a934fc4bd4343799f6f5655d07ef25208ed0be5a&vid=a934fc4bd4343799f6f5655d07ef25208ed0be5a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR