2-Methoxyethyl-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methoxyethyl 4-methylbenzenesulfonate, 2-Methoxyethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-Methoxyethyl Ester, p-Toluenesulfonic acid 2-Methoxyethyl Ester

T1303-25G T1303-500G T1303-100G
TCT1303-025GEA 161 CAD
TCT1303-025G TCT1303-500G TCT1303-100G
2-Methoxyethyl-p-toluenesulfonate ≥98.0%
2-Methoxyethyl-p-toluenesulfonate

CAS Number: 17178-10-8
MDL Number: MFCD00025910
Molecular Formula: C10H14O4S
Molecular Weight: 230.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 141
Melting point (°C): 10
Specific Gravity (20/20): 1.21


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid 2-Methoxyethyl Ester

Formula: C₁₀H₁₄O₄S
MW: 230.28 g/mol
Boiling Pt: 141 °C
Melting Pt: 10 °C
Density: 1.21 (20 °C)
MDL Number: MFCD00025910
CAS Number: 17178-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec821fcf0d8fa9c842a7a89265a53d80af10bfc6&vid=ec821fcf0d8fa9c842a7a89265a53d80af10bfc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR