meso-Tetra(4-carboxyphenyl)porphine ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Tetrakis(4-carboxyphenyl)porphyrin, 4,4,4,4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), TCPP, Tetrakis (4-carboxyphenyl) porphyrin
[Ultra-high sensitive spectrophotometric reagent for Cu, Cd][For the simultaneous determination of metals by HPLC]
CAS Number: 14609-54-2
MDL Number: MFCD00064860
Molecular Formula: C48H30N4O8
Molecular Weight: 790.79
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Product Categories: Bioscience, Porphyrins, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI), Porphines (Chelating Agents)
Other Product Names / Synonyms: Tetrakis(4-carboxyphenyl)porphyrin
Formula:
C₄₈H₃₀N₄O₈ MW: 790.79 g/mol |
MDL Number:
MFCD00064860 CAS Number: 14609-54-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...