Triethanolamine borate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2',2''-Nitrilotriethyl Borate

T1291-25G T1291-500G T1291-100G
TCT1291-025GEA 60.2 CAD
TCT1291-025G TCT1291-500G TCT1291-100G
Triethanolamine borate ≥95.0% (by titrimetric analysis)
Triethanolamine borate

CAS Number: 283-56-7
MDL Number: MFCD00003272
Molecular Formula: C6H12BNO3
Molecular Weight: 156.98
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 237


Other Product Names / Synonyms: 2,2',2''-Nitrilotriethyl Borate

Formula: C₆H₁₂BNO₃
MW: 156.98 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00003272
CAS Number: 283-56-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79fcb09037d38521b129d2c064dae5e0035b4c29&vid=79fcb09037d38521b129d2c064dae5e0035b4c29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR