Tetrabutylammonium thiocyanate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T1278-250G T1278-25G
TCT1278-250GEA 867.74 CAD
TCT1278-250G TCT1278-025G
Tetrabutylammonium thiocyanate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium thiocyanate

CAS Number: 3674-54-2
MDL Number: MFCD00012140
Molecular Formula: C17H36N2S
Molecular Weight: 300.55
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 123


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Tetrabutylammonium Rhodanide

Formula: C₁₇H₃₆N₂S
Melting Pt: 123 °C
MDL Number: MFCD00012140
CAS Number: 3674-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67ef5b35af7635b63fd292ae03f78ff4125af9c6&vid=67ef5b35af7635b63fd292ae03f78ff4125af9c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR