TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate] ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: alpha,beta,gamma,delta-Tetrakis(1-methylpyridinium-4-yl)porphyrin p-Toluenesulfonate, TMPyP4 tosylate, alpha,beta,gamma,delta-Tetrakis(1-methylpyridinium-4-yl)porphyrin p-Toluenesulphonate, 5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulphonate), 5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate)

A5014-1G A5014-100MG
TCA5014-001GEA 504.02 CAD
TCA5014-001G TCA5014-100MG
TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate] ≥98.0% (by total nitrogen basis)
TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate]

[Ultra-high sensitive spectrophotometric reagent for Cu, Mg][For the simultaneous determination of metals by HPLC]
CAS Number: 36951-72-1
MDL Number: MFCD00013468
Molecular Formula: C72H66N8O12S4
Molecular Weight: 1363.60
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Porphyrins, Cancer Research Reagents, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI), Porphines (Chelating Agents)
Other Product Names / Synonyms: alpha,beta,gamma,delta-Tetrakis(1-methylpyridinium-4-yl)porphyrin p-Toluenesulfonate

Formula: C₇₂H₇₄N₈O₁₂S₄
MW: 1371.66 g/mol
MDL Number: MFCD00013468
CAS Number: 36951-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=885fd963654539abfa12d4e7df444dd8dca514bf&vid=885fd963654539abfa12d4e7df444dd8dca514bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR