Tetrabutylammonium triiodide ≥96.0% (by titrimetric analysis)

Supplier: TCI America

T1271-25G T1271-5G
TCT1271-025GEA 325.48 CAD
TCT1271-025G TCT1271-5G
Tetrabutylammonium triiodide ≥96.0% (by titrimetric analysis)
Tetrabutylammonium triiodide

CAS Number: 13311-45-0
MDL Number: MFCD00059123
Molecular Formula: C16H36I3N
Molecular Weight: 623.18
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: Purple
Melting point (°C): 71

Formula: C₁₆H₃₆I₃N
Melting Pt: 71 °C
MDL Number: MFCD00059123
CAS Number: 13311-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=facbea535a0a65e5309eb3ab7252c5ffa9b17a87&vid=facbea535a0a65e5309eb3ab7252c5ffa9b17a87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR