Tetrabutylammonium dibromoiodide ≥97.0%

Supplier: TCI America

Synonyms: Tetrabutylammonium Dibromoiodide

T1269-10G T1269-1G
TCT1269-010GEA 587.76 CAD
TCT1269-010G TCT1269-001G
Tetrabutylammonium dibromoiodide ≥97.0%
Tetrabutylammonium dibromoiodide

CAS Number: 15802-00-3
MDL Number: MFCD00059121
Molecular Formula: C16H36Br2IN
Molecular Weight: 529.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 81

Formula: C₁₆H₃₆Br₂IN
Melting Pt: 81 °C
MDL Number: MFCD00059121
CAS Number: 15802-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bd51674e35f48f0e1d996a14969965f18334253&vid=3bd51674e35f48f0e1d996a14969965f18334253&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR