(3-Chloropropyl)triethoxysilane ≥97.0%

Supplier: TCI America

Synonyms: 3-Chloropropyltriethoxysilane, Triethoxy(3-chloropropyl)silane, 3-(Triethoxysilyl)propylchloride

T1253-500ML T1253-25ML T1253-100ML
TCT1253-500MLEA 105.86 CAD
TCT1253-500ML TCT1253-025ML TCT1253-100ML
(3-Chloropropyl)triethoxysilane ≥97.0%
(3-Chloropropyl)triethoxysilane

CAS Number: 5089-70-3
MDL Number: MFCD00018985
Molecular Formula: C9H21ClO3Si
Molecular Weight: 240.80
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 102
Flash Point (°C): 86
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Triethoxysilyl)propyl Chloride

Formula: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
MW: 240.8 g/mol
Boiling Pt: 102 °C
Density: 1.01 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00018985
CAS Number: 5089-70-3
EINECS: 225-805-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a256c97a32b08ff1c428f758a4da4261ef8ebf35&vid=a256c97a32b08ff1c428f758a4da4261ef8ebf35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR