Ethyl L-(-)-O-tosyllactate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1241-1G T1241-25G
TCT1241-001GEA 45.82 CAD
TCT1241-001G TCT1241-25G
Ethyl L-(-)-O-tosyllactate ≥98.0% (by titrimetric analysis)
Ethyl L-(-)-O-tosyllactate

CAS Number: 57057-80-4
MDL Number: MFCD06809725
Molecular Formula: C12H16O5S
Molecular Weight: 272.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Freezing point (°C): 32
Specific rotation [a]20/D: -36.5 deg (C=5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Building Blocks
Other Product Names / Synonyms: Ethyl L-(-)-O-(p-Toluenesulfonyl)lactate, L-(-)-O-(p-Toluenesulfonyl)lactic Acid Ethyl Ester, L-(-)-O-Tosyllactic Acid Ethyl Ester

Formula: C₁₂H₁₆O₅S
MDL Number: MFCD06809725
CAS Number: 57057-80-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b818730ed348c99a1d5e5ab1681fa250eb7b01&vid=f9b818730ed348c99a1d5e5ab1681fa250eb7b01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR