Tetramethylammonium sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1240-25G T1240-5G
TCT1240-025GEA 285.98 CAD
TCT1240-025G TCT1240-005G
Tetramethylammonium sulfate ≥98.0% (by titrimetric analysis)
Tetramethylammonium sulfate

CAS Number: 14190-16-0
MDL Number: MFCD00012139
Molecular Formula: C8H24N2O4S
Molecular Weight: 244.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 285

Formula: C₈H₂₄N₂O₄S
Melting Pt: 285 °C
MDL Number: MFCD00012139
CAS Number: 14190-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51ed96d47d168eab8374482f879537738ad19cab&vid=51ed96d47d168eab8374482f879537738ad19cab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR