Stilbazo

Supplier: TCI America

A5011-1G A5011-5G
TCA5011-001GEA 121.66 CAD
TCA5011-001G TCA5011-005G
Stilbazo
Stilbazo

[Spectrophotometric reagent for Al and other metals]
CAS Number: 1571-36-4
MDL Number: MFCD00045752
Molecular Formula: C26H26N6O10S2
Molecular Weight: 646.65
Form: Crystal
Color: Deep Yellow Red


Other Product Names / Synonyms: Stilbene-4,4'-bis[(1-azo)-3,4-dihydroxybenzene]-2,2'-disulfonic Acid Diammonium Salt

Formula: C₂₆H₂₆N₆O₁₀S₂
MDL Number: MFCD00045752
CAS Number: 1571-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8096ea3cb94919a5bf4b24c6f4db7c8fd6cb9ba3&vid=8096ea3cb94919a5bf4b24c6f4db7c8fd6cb9ba3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR