2,2',4,4'-Tetramethylbenzophenone ≥96.0%

Supplier: TCI America

T1232-25G T1232-5G
TCT1232-025GEA 477.16 CAD
TCT1232-025G 76220-494
2,2',4,4'-Tetramethylbenzophenone ≥96.0%
2,2',4,4'-Tetramethylbenzophenone

CAS Number: 3478-88-4
MDL Number: MFCD00060094
Molecular Formula: C17H18O
Molecular Weight: 238.33
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 188
Specific Gravity (20/20): 1.04

Formula: C₁₇H₁₈O
Boiling Pt: 188 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00060094
CAS Number: 3478-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17a1ed30ba9e497d8356157130ed5b51d8661d9b&vid=17a1ed30ba9e497d8356157130ed5b51d8661d9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR