Trimellitic acid trimethyl ester ≥95.0%

Supplier: TCI America

T1229-500G T1229-25G
TCT1229-500GEA 235.42 CAD
TCT1229-500G TCT1229-025G
Trimellitic acid trimethyl ester ≥95.0%
Trimellitic acid trimethyl ester

CAS Number: 2459-10-1
MDL Number: MFCD00043629
Molecular Formula: C12H12O6
Molecular Weight: 252.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 189
Flash Point (°C): 182
Specific Gravity (20/20): 1.26


Other Product Names / Synonyms: 1,2,4-Benzenetricarboxylic Acid Trimethyl Ester, Trimellitic Acid Trimethyl Ester, Trimethyl 1,2,4-Benzenetricarboxylate

Formula: C₁₂H₁₂O₆
Boiling Pt: 189 °C
Density: 1.26 (20 °C)
Flash Pt: 182 °C
MDL Number: MFCD00043629
CAS Number: 2459-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22d11f06f875a526fa1e2029e20984d9c8a54783&vid=22d11f06f875a526fa1e2029e20984d9c8a54783&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR