2-(p-Tolyl)pyridine ≥97.0%
Supplier: TCI America
T1228-25G
T1228-5G
TCT1228-025GEA
170.64
CAD
TCT1228-025G
TCT1228-005G
2-(p-Tolyl)pyridine ≥97.0%
2-(p-Tolyl)pyridine
CAS Number: 4467-06-5
MDL Number: MFCD00006281
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 157
Flash Point (°C): 110
Specific Gravity (20/20): 1.06
Formula:
C₁₂H₁₁N Boiling Pt: 157 °C Density: 1.06 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006281 CAS Number: 4467-06-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...