(R)-(-)-2-Oxothiazolidine-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(R)-2-Oxothiazolidine-4-carboxylic acid, Procysteine
T1223-1G
T1223-5G
TCT1223-001GEA
64.78
CAD
TCT1223-001G
TCT1223-005G
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
CAS Number: 19771-63-2
MDL Number: MFCD00066092
Molecular Formula: C4H5NO3S
Molecular Weight: 147.15
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -64 deg (C=1, H2O)
Other Product Names / Synonyms: L-2-Oxothiazolidine-4-carboxylic Acid
Formula:
C₄H₅NO₃S MW: 147.15 g/mol |
MDL Number:
MFCD00066092 CAS Number: 19771-63-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...