Sulfonazo III sodium salt ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
2,7-Bis(2-sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonic Acid Tetrasodium Salt, 2,7-Bis(2-sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic Acid Tetrasodium Salt
[Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba]
CAS Number: 68504-35-8
MDL Number: MFCD00003942
Molecular Formula: C22H16N4O14S4
Molecular Weight: 776.55
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 2,7-Bis(2-sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonic Acid Tetrasodium Salt
Formula:
C₂₂H₁₆N₄O₁₄S₄ MW: 776.55 g/mol |
MDL Number:
MFCD00003942 CAS Number: 68504-35-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...