22-Tricosenoic acid ≥97.0%

Supplier: TCI America

T1211-100MG T1211-1G
TCT1211-100MGEA 210.14 CAD
TCT1211-100MG TCT1211-001G
22-Tricosenoic acid ≥97.0%
22-Tricosenoic acid

CAS Number: 65119-95-1
MDL Number: MFCD00060118
Molecular Formula: C23H44O2
Molecular Weight: 352.60
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 76

Formula: C₂₃H₄₄O₂
MW: 352.60 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00060118
CAS Number: 65119-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f08ec8ace4d25f91686744bc14d1ba55e069d58d&vid=f08ec8ace4d25f91686744bc14d1ba55e069d58d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR