Dimethylsulfonazo III
Supplier: TCI America
Synonyms:
2,7-Bis(2-sulfo-p-tolylazo)-1,8-dihydroxy-3,6-naphthalenedisulfonic acid, 2,7-Bis(2-sulpho-p-tolylazo)-1,8-dihydroxy-3,6-naphthalenedisulphonic acid, 2,7-Bis[(4-methyl-2-sulphophenyl)azo]-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,7-Bis[(4-methyl-2-sulfophenyl)azo]-1,8-dihydroxynaphthalene-3,6-disulfonic acid
[Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba]
CAS Number: 14979-11-4
MDL Number: MFCD00151118
Molecular Formula: C24H20N4O14S4
Molecular Weight: 716.68
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: 2,7-Bis[(4-methyl-2-sulfophenyl)azo]-1,8-dihydroxynaphthalene-3,6-disulfonic Acid, 2,7-Bis(2-sulfo-p-tolylazo)-1,8-dihydroxy-3,6-naphthalenedisulfonic Acid
Formula:
C₂₄H₂₀N₄O₁₄S₄ MW: 716.68 g/mol |
MDL Number:
MFCD00151118 CAS Number: 14979-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...