5,6,7,8-Tetrahydroquinoline ≥96.0% (by GC, titration analysis)
Supplier: TCI America
T1150-100ML
T1150-10ML
TCT1150-100MLEA
759.98
CAD
TCT1150-100ML
TCT1150-010ML
5,6,7,8-Tetrahydroquinoline ≥96.0% (by GC, titration analysis)
5,6,7,8-Tetrahydroquinoline
CAS Number: 10500-57-9
MDL Number: MFCD00006734
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 222
Flash Point (°C): 86
Specific Gravity (20/20): 1.03
Formula:
C₉H₁₁N Boiling Pt: 222 °C Density: 1.03 (20 °C) Flash Pt: 86 °C |
MDL Number:
MFCD00006734 CAS Number: 10500-57-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...